missing translation for 'onlineSavingsMsg'
Learn More

Briciclib, MedChemExpress

Product Code. 16497038
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497038 10 mg 10mg
16493087 50 mg 50mg
16449587 5 mg 5mg
16447096 100 mg 100mg
16441609 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497038 Supplier Medchem Express Supplier No. HY16366
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Briciclib (ON 014185) is a derivative of ON 013100, and has the potential in targeting eIF4E for solid cancers.

Briciclib (ON 014185) is a derivative of ON 013100, and has the potential in targeting eIF4E for solid cancers.

Specifications

Chemical Name or Material Briciclib
CAS 865783-99-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H23O10PS
Quantity 10 mg
Synonym ON 014185
Solubility Information DMSO : ≥ 31 mg/mL (65.34 mM)
SMILES O=S(CC1=CC(OP(O)(O)=O)=C(OC)C=C1)(/C=C/C2=C(OC)C=C(OC)C=C2OC)=O
Molecular Weight (g/mol) 474.42
Formula Weight 474.42
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.