missing translation for 'onlineSavingsMsg'
Learn More

Brij™ C20

Product Code. 10748562 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Unit Size:
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10748562

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-95-9 | C56H114O21 | 1123.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-95-9
Molecular Formula C56H114O21
Molecular Weight (g/mol) 1123.51
MDL Number MFCD00080892
InChI Key NLMKTBGFQGKQEV-UHFFFAOYSA-N
Synonym Polyoxyethylene(20) cetyl ether
PubChem CID 2724259
SMILES CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White
Acid Value 1mg KOH/g max.
Physical Form Pellets
Melting Point 35.4°C to 39.6°C
Quantity 500 g
Flash Point >149°C
Hydroxyl Value 45 to 60mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C16H33(OCH2CH2)20OH
Water 1% max.
Merck Index 10, 7449
Solubility Information Solubility in water: soluble. Other solubilities: soluble in lower alcohols, insoluble in propylene glycol, coal-tar hydrocarbo, n, cottonseed and mineral oil
IUPAC Name 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
Formula Weight 1123.5
Chemical Name or Material Brij™ C20
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.