missing translation for 'onlineSavingsMsg'
Learn More

Brilacidin, MedChemExpress

Product Code. 16461889
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16461889

Brand: Medchem Express HY19892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brilacidin (PMX 30063) is an anti-infective antimicrobial with MIC90s of 1 and 8 μg/mL for Gram-positive bacteria Streptococcus pneumonia and Streptococcus viridans, and MIC90 of 8 and 4 μg/mL for Gram-negative bacteria Haemophilus influenza and Pseudomonas aeruginosa. Brilacidin is a defensin mimetic antibiotic compound.

Brilacidin (PMX 30063) is an anti-infective antimicrobial with MIC90s of 1 and 8 μg/mL for Gram-positive bacteria Streptococcus pneumonia and Streptococcus viridans, and MIC90 of 8 and 4 μg/mL for Gram-negative bacteria Haemophilus influenza and Pseudomonas aeruginosa. Brilacidin is a defensin mimetic antibiotic compound.

Specifications

Chemical Name or Material Brilacidin
Physical Form Solid
CAS 1224095-98-0
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C40H50F6N14O6
For Use With (Application) COVID-19-immunoregulation
Synonym PMX 30063
SMILES O=C(C1=NC=NC(C(NC2=CC(C(F)(F)F)=CC(NC(CCCCNC(N)=N)=O)=C2O[C@H]3CNCC3)=O)=C1)NC4=CC(C(F)(F)F)=CC(NC(CCCCNC(N)=N)=O)=C4O[C@H]5CNCC5
Molecular Weight (g/mol) 936.91
Formula Weight 936.91
Percent Purity 92.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.