missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ ESBL Agar

Product Code. 11974102
Change view
Click to view available options
Format:
90mm Monoplate
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
11974102 90mm Monoplate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11974102 Supplier Thermo Scientific™ Supplier No. PO5302A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Screen clinical samples for the presence of extended-spectrum beta-lactamase (ESBL)-producing bacteria on this novel formulation which uses color to clearly differentiate E.coli.

Detect extended-spectrum beta-lactamase-producing Enterobacteriaceae while selectively inhibiting AmpC resistance mechanisms using Thermo Scientific™ Brilliance™ ESBL Agar. Improve processing time through the direct inoculation from screening swab, isolated colony or from a liquid suspension.

ESBL-producing E. coli grow as either blue or pink colonies. ESBL-producing members of the KESC group produce green colonies; Proteus, Morganella and Providencia do not utilize either chromogen but are able to deaminate tryptohan, resulting in tan colonies with a brown halo.

Brilliance ESBL Agar is a selective medium for the screening of clinical samples for the presence of extended-spectrum beta-lactamase (ESBL) producing bacteria. Easily and clearly differentiate E.coli and the KESC group by different colors to help to identify ESBL producing organisms . The inhibition of AmpC producers reduce false positives and the need for confirmatory tests.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Oyster white, opaque
Content And Storage 2° to 12°C, in the dark
Description Brilliance ESBL Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.