missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Blue FCF (Technical Grade), TRC

Product Code. 30005814
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005814 5 g 5g
30005848 10 g 10g
30011041 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005814 Supplier Toronto Research Chemicals Supplier No. TRCB6774805G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37 H34 N2 O9 S3 . 2 Na
Molecular Weight (g/mol) 792.85
Synonym Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-, hydroxide, inner salt, disodium salt, Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-, inner salt, disodium salt (9CI), Brilliant Blue FCF (6CI), C.I. Acid Blue 9, disodium salt (7CI,8CI), AB 9, AB 9 (dye), Blue 1, Blue 1206, Blue FCF, Blue No. 1, Brilliant Blue, Brilliant Blue E 133, Brilliant Blue FCF Supra, C.I. 42090, C.I. Food Blue 2, Canacert Brilliant Blue FCF, D and C Blue No. 1, D&C Blue No. 1, Disodium erioglaucine, Dolkwal Brilliant Blue, Duasyn Acid Blue AE 02, E 133, E 133 (dye), Erioglaucine disodium salt, FD and C Blue 1, FD and C Blue No. 1, FD&C Blue No. 1, FDC Blue 1, Food Blue 1, Food Blue 2, Food Blue Dye No. 1, Food Blue No. 1, Food Blue No. 2, Food Brilliant blue, Hexacol Brilliant Blue A, Japan Blue 1, Japan Blue No. 1, Japan Food Blue 1, Japan Food Blue No. 1, K-RCP-B 1AL, Kayaset Blue J-FG, Puricolor Blue ABL 9, Sicovit Brilliant Blue 85E133, Unicert Blue O 5601J, Usacert Blue No. 1, Usacert FD and C Blue No. 1
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMILES [Na+].[Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccccc5S(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C37H36N2O9S3.2Na/c1-3-38(25-27-9-7-11-33(23-27)49(40,41)42)31-19-15-29(16-20-31)37(35-13-5-6-14-36(35)51(46,47)48)30-17-21-32(22-18-30)39(4-2)26-28-10-8-12-34(24-28)50(43,44)45;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/p-2
Formula Weight 792.1222
Chemical Name or Material Brilliant Blue FCF (E133)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.