missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Crocein

Product Code. 11457596
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11457596 50 g 50g
11467596 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 11457596

Brand: Thermo Scientific Alfa Aesar A14642.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5413-75-2 | C22H16N4Na2O7S2 | 558.491 g/mol

Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5413-75-2
Molecular Formula C22H16N4Na2O7S2
Molecular Weight (g/mol) 558.491
MDL Number MFCD00003907
InChI Key BMBHMIGLJQAUTJ-HKWNQDJZSA-N
Synonym C.I. 27290; Crocein Scarlet MOO
PubChem CID 88641610
IUPAC Name (8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Solubility Information Soluble in water
Formula Weight 556.49
Quantity 50 g
Chemical Name or Material Brilliant Crocein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.