missing translation for 'onlineSavingsMsg'
Learn More

Brimonidine tartrate, MedChemExpress

Product Code. 16478406
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478406 100 mg 100mg
16484437 50 mg 50mg
16468568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478406 Supplier Medchem Express Supplier No. HYB0659A100mg
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Brimonidine tartrate (UK 14304 tartrate) is a full α2-adrenergic receptor (α2-AR) agonist.

Brimonidine tartrate (UK 14304 tartrate) is a full α2-adrenergic receptor (α2-AR) agonist.

Specifications

Chemical Name or Material Brimonidine tartrate
CAS 70359-46-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H16BrN5O6
Quantity 100 mg
Synonym UK 14304 tartrateAGN190342 tartrate
Solubility Information DMSO : 50 mg/mL (113.07 mM; Need ultrasonic) ∣H2O : 25 mg/mL (56.53 mM; Need ultrasonic)
SMILES O=C(O)[C@H](O)[C@@H](O)C(O)=O.BrC1=C2N=CC=NC2=CC=C1NC3=NCCN3
Molecular Weight (g/mol) 442.22
Formula Weight 442.22
Percent Purity 99.19%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.