missing translation for 'onlineSavingsMsg'
Learn More

Brinzolamide, MedChemExpress

Product Code. 16478376
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478376 100 mg 100mg
16448538 10 mg 10mg
16424407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478376 Supplier Medchem Express Supplier No. HYB0588
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor with anIC50 value of 3.2 nM. Brinzolamide hydrochloride reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion. Brinzolamide can be used in glaucoma disease research.

Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor with anIC50 value of 3.2 nM. Brinzolamide hydrochloride reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion. Brinzolamide can be used in glaucoma disease research.

Specifications

Chemical Name or Material Brinzolamide
CAS 138890-62-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H21N3O5S3
Quantity 100 mg
Synonym AL-4862
Solubility Information DMSO : 100 mg/mL (260.75 mM; Need ultrasonic)
SMILES O=S(C(S1)=CC2=C1S(N(CCCOC)C[C@@H]2NCC)(=O)=O)(N)=O
Molecular Weight (g/mol) 383.51
Formula Weight 383.51
Percent Purity 99.33%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.