missing translation for 'onlineSavingsMsg'
Learn More

Brivudine, MedChemExpress

Product Code. 16451917
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451917 50 mg 50mg
16471086 200 mg 200mg
16423376 25 mg 25mg
16426236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451917 Supplier Medchem Express Supplier No. HY13578
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Brivudine is a thymidine analogue with antiviral activity, indicated for the early treatment of acute herpes zoster.

Brivudine is a thymidine analogue with antiviral activity, indicated for the early treatment of acute herpes zoster.

Specifications

Chemical Name or Material Brivudine
Physical Form Solid
CAS 69304-47-8
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H13BrN2O5
For Use With (Application) COVID-19-anti-virus
Synonym Bromovinyldeoxyuridine BVDU
Solubility Information DMSO : 200 mg/mL (600.35 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(/C=C/Br)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 333.14
Formula Weight 333.14
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.