missing translation for 'onlineSavingsMsg'
Learn More

Broflanilide-desmethyl, Dr. Ehrenstorfer

Product Code. 30318541
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318541 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318541 Supplier Dr. Ehrenstorfer Supplier No. DREC10668010
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Broflanilide-desmethyl
CAS 1207977-84-1
Analyte or Component Names Broflanilide-desmethyl
Shipping Condition +20°C
Molecular Formula C24 H12 Br F11 N2 O2
Certifications/Compliance ISO/IEC 17025
Synonym 3-(Benzoylamino)-N-[2-bromo-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]-6-(trifluoromethyl)phenyl]-2-fluorobenzamide
InChI Formula InChI=1S/C24H12BrF11N2O2/c25-15-10-12(21(27,23(31,32)33)24(34,35)36)9-14(22(28,29)30)18(15)38-20(40)13-7-4-8-16(17(13)26)37-19(39)11-5-2-1-3-6-11/h1-10H,(H,37,39)(H,38,40)
SMILES Fc1c(NC(=O)c2ccccc2)cccc1C(=O)Nc3c(Br)cc(cc3C(F)(F)F)C(F)(C(F)(F)F)C(F)(F)F
IUPAC Name 3-benzamido-N-[2-bromo-4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-6-(trifluoromethyl)phenyl]-2-fluorobenzamide
Molecular Weight (g/mol) 649.2507
Quantity 10 mg
Formula Weight 647.9907 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.