missing translation for 'onlineSavingsMsg'
Learn More

Brombuterol Hydrochloride, TRC

Product Code. 30011165
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011165 500 mg 500mg
30010586 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011165 Supplier Toronto Research Chemicals Supplier No. TRCB678535500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21912-49-2
Molecular Formula C12 H18 Br2 N2 O . Cl H
Molecular Weight (g/mol) 402.55
Synonym Benzenemethanol, 4-amino-3,5-dibromo-α-[[(1,1-dimethylethyl)amino]methyl]-, monohydrochloride (9CI), Benzyl alcohol, 4-amino-3,5-dibromo-α-[(tert-butylamino)methyl]-, monohydrochloride (8CI), 4-Amino-3,5-dibromo-α-[(tert-butylamino)methyl]benzyl alcohol hydrochloride
IUPAC Name 1-(4-amino-3,5-dibromophenyl)-2-(tert-butylamino)ethanol;hydrochloride
SMILES Cl.CC(C)(C)NCC(O)c1cc(Br)c(N)c(Br)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H18Br2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H
Formula Weight 399.9553
Percent Purity >95%
Chemical Name or Material Brombuterol hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.