missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine hydrochloride, MedChemExpress

Product Code. 16460898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460898 5 g 5g
16448458 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460898 Supplier Medchem Express Supplier No. HYB0372A
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bromhexine hydrochloride is a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM. Bromhexine hydrochloride can prevent and manage SARS-CoV-2 infection. Bromhexine hydrochloride is an autophagy agonist. Bromhexine hydrochloride is a mucolytic cough suppressant and has the potential for a range of respiratory conditions.

Bromhexine hydrochloride is a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM. Bromhexine hydrochloride can prevent and manage SARS-CoV-2 infection. Bromhexine hydrochloride is an autophagy agonist. Bromhexine hydrochloride is a mucolytic cough suppressant and has the potential for a range of respiratory conditions.

Specifications

Chemical Name or Material Bromhexine hydrochloride
CAS 611-75-6
Color White
Purity Grade Notes Research
Molecular Formula C14H21Br2ClN2
Quantity 5 g
Solubility Information DMSO : 20 mg/mL (48.47 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (8.07 mM; Need ultrasonic)
SMILES CN(C1CCCCC1)CC2=CC(Br)=CC(Br)=C2N.Cl
Molecular Weight (g/mol) 412.59
Formula Weight 412.59
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.