missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine Hydrochloride, TRC

Product Code. 30138742
Change view
Click to view available options
Quantity:
50 mg
5 g
50 g
Unit Size:
50g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138742 5 g 5g
30008982 50 g 50g
17999564 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138742 Supplier Toronto Research Chemicals Supplier No. TRCB6786005G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-75-6
Molecular Formula C14 H20 Br2 N2 . Cl H
Molecular Weight (g/mol) 412.59
Synonym Benzenemethanamine, 2-amino-3,5-dibromo-N-cyclohexyl-N-methyl-, hydrochloride (1:1), Benzenemethanamine, 2-amino-3,5-dibromo-N-cyclohexyl-N-methyl-, monohydrochloride (9CI), Toluene-α,2-diamine, 3,5-dibromo-Nα-cyclohexyl-Nα-methyl-, monohydrochloride (8CI), 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine monohydrochloride, Auxit, Bisolvon, Bisolvon hydrochloride, Bromhexine chloride, Bromhexine hydrochloride, Bromhexine monohydrochloride, Cyclohexylmethyl(2-amino-3,5-dibromobenzyl)ammonium chloride, N-Cyclohexyl-N-methyl-(2-amino-3,5-dibromobenzyl)ammonium chloride, N-Cyclohexyl-N-methyl-N-(2-amino-3,5-dibromobenzyl)ammonium chloride, Nα-Cyclohexyl-Nα-methyl-3,5-dibromotoluene-α,2-diamine hydrochloride, Ophtosol, Quentan
IUPAC Name 2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]aniline;hydrochloride
SMILES Cl.CN(Cc1cc(Br)cc(Br)c1N)C2CCCCC2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H20Br2N2.ClH/c1-18(12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17;/h7-8,12H,2-6,9,17H2,1H3;1H
Formula Weight 409.976
Percent Purity >95%
Chemical Name or Material Bromhexine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.