missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromide Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical A 008 500
Code : 04
Additional Details : Weight : 0.50000kg
Specifications
Bromide | |
Bottle | |
Single-component Standard Solution | |
H2O |
1g/L | |
500 mL | |
Br- | |
Liquid |