missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 1M solution in acetic acid, AcroSeal™

Product Code. 10530364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10530364 100 mL AcroSeal™ Glass bottle 100mL
10445465 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10530364 Supplier Thermo Scientific Acros Supplier No. 388401000
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    108.00 € / 100mL
    Save 8.00 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 7726-95-6 | Br | 79.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromine
Color Red
Name Note 1M solution in acetic acid
CAS 7726-95-6, 64-19-7
Concentration 0.90 to 1.10M
Density 3.111
Packaging AcroSeal™ Glass bottle
Molecular Formula Br
Linear Formula Br2
MDL Number MFCD00010896
Quantity 100 mL
Flash Point 40°C
Merck Index 15, 1401
Physical Form Liquid
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
Solubility Information Solubility in water: soluble.
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES [Br-]
IUPAC Name bromide
Molecular Weight (g/mol) 79.91
PubChem CID 24408
ChEBI CHEBI:29224
Formula Weight 159.82
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine, 1M solution in acetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.