missing translation for 'onlineSavingsMsg'
Learn More

α-Bromo-2,5-difluorotoluene, 98%, ACROS Organics™

Product Code. 10195460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
Product Code. Quantity Packaging unitSize
10195460 10 g Glass bottle 10g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10195460

Brand: Thermo Scientific Acros 311550100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 85117-99-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H5BrF2
Linear Formula FC6H3CHBr
Refractive Index 1.5246 to 1.5266 (20°C, 589nm)
MDL Number MFCD00009897
Quantity 10 g
Synonym 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i
InChI Key ONWGSWNHQZYCFK-UHFFFAOYSA-N
SMILES FC1=CC=C(F)C(CBr)=C1
IUPAC Name 2-(bromomethyl)-1,4-difluorobenzene
Molecular Weight (g/mol) 207.02
PubChem CID 522830
Formula Weight 207.02
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material α-Bromo-2, 5-difluorotoluene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.