missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-m-tolunitrile, 95%

Product Code. p-3777729 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10091940

Brand: Thermo Scientific Acros 163510100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28188-41-2 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28188-41-2
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001809
InChI Key CVKOOKPNCVYHNY-UHFFFAOYSA-N
Synonym 3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile
PubChem CID 97249
IUPAC Name 3-(bromomethyl)benzonitrile
SMILES BrCC1=CC=CC(=C1)C#N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 92°C to 97°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CN
Quantity 10 g
Beilstein 09, II, 326
Formula Weight 196.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material α-Bromo-m-tolunitrile
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromo-m-tolunitrile
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.