missing translation for 'onlineSavingsMsg'
Learn More

alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%

Product Code. 10351194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351194 1 g Glass bottle 1g
10740781 5 g Glass bottle 5g
10678444 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351194 Supplier Thermo Scientific Acros Supplier No. 312990010
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 402-49-3 | C8H6BrF3 | 239.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-49-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD00000403
InChI Key IKSNDOVDVVPSMA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
PubChem CID 123062
IUPAC Name 1-(bromomethyl)-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1CBr)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 30°C to 34°C
Color White to Yellow
Density 1.5400g/mL
Boiling Point 65°C to 69°C (5.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CH2Br
Quantity 1 g
Specific Gravity 1.54
Formula Weight 239.04
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material α'-Bromo-α, α, α-trifluoro-p-xylene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.