missing translation for 'onlineSavingsMsg'
Learn More

Bromohydroquinone, 94%, pract., Thermo Scientific™

Product Code. 10370194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10370194 50 g Glass bottle 50g
10239450 1 g Glass bottle 1g
10569272 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10370194 Supplier Thermo Scientific Acros Supplier No. 403040500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-69-7
MDL Number MFCD00041747
InChI Key REFDOIWRJDGBHY-UHFFFAOYSA-N
Synonym bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
PubChem CID 68502
IUPAC Name 2-bromobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Br)O

Specifications

Melting Point 104°C to 110°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Molecular Formula C6H5BrO2
Quantity 50 g
Molecular Weight (g/mol) 189.008
Formula Weight 189.01
Percent Purity 39.7 to 42.7% (bromine, by combustion), Separat. Tech. GC ≥88%
Grade Practical
Physical Form Fine Crystalline Powder
Chemical Name or Material Bromohydroquinone
Show More Show Less
compliance-icons
Product Identifier
  • Bromohydroquinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.