missing translation for 'onlineSavingsMsg'
Learn More

Bromopentafluorobenzene, 99%

Product Code. 10070860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070860 5 mL Glass bottle 5mL
10413585 25 mL Glass bottle 25mL
10443585 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10070860 Supplier Thermo Scientific Acros Supplier No. 107220050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 344-04-7 | C6BrF5 | 246.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-04-7
Molecular Formula C6BrF5
Molecular Weight (g/mol) 246.96
MDL Number MFCD00000287
InChI Key XEKTVXADUPBFOA-UHFFFAOYSA-N
Synonym bromopentafluorobenzene, pentafluorobromobenzene, benzene, bromopentafluoro, bromoperfluorobenzene, pentafluorophenyl bromide, 2,3,4,5,6-pentafluorobromobenzene, benzene, 1-bromo-2,3,4,5,6-pentafluoro, 1-bromo-pentafluorobenzene, 2,3,4,5,6-bromopentafluorobenzene, 1-bromo-2,3,4,5,6-pentafluoro-benzene
PubChem CID 9578
IUPAC Name 1-bromo-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point -31°C
Color Colorless
Density 1.9810g/mL
Boiling Point 137°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6F5
Refractive Index 1.4485 to 1.4505
Quantity 5 mL
Specific Gravity 1.981
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,benzene
Formula Weight 246.96
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bromopentafluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • Bromopentafluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.