missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromostyrene, 95%, stabilized

Product Code. 10507963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10507963 5 g Glass bottle 5g
10662613 25 g Glass bottle 25g
10095153 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10507963 Supplier Thermo Scientific Acros Supplier No. 365800050
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-81-7 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-81-7
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00012229
InChI Key SRXJYTZCORKVNA-UHFFFAOYSA-N
Synonym 1-bromovinyl benzene, alpha-bromostyrene, benzene, 1-bromoethenyl, 1-bromostyrene, styrene, alpha-bromo, 1-phenylvinyl bromide, alpha-bromostyrol russian, styrene, .alpha.-bromo, unii-lf0sj1821n, alpha-bromostyrol
PubChem CID 66828
IUPAC Name 1-bromoethenylbenzene
SMILES BrC(=C)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -44°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 67°C to 70°C (4.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(Br)=CH2
Refractive Index 1.5870 to 1.589
Quantity 5 g
Specific Gravity 1.41
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Stabilizer about 0.05% 3, 5-di-tert-butylcatechol
Formula Weight 183.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material α-Bromostyrene, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromostyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.