missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol blue sodium salt, MP Biomedicals™

Product Code. 11447801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447801 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447801 Supplier MP Biomedicals Supplier No. 0215052405
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 34722-90-2
Color Green
Molecular Formula C27H27Br2NaO5S
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
Quantity 5 g
Synonym cfn0fbc3yh, unii-cfn0fbc3yh, sodium bromothymol blue, bromothymol blue sodium salt, 3',3-dibromothymolsulfonephthalein sodium salt, bromthymol blue sodium, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-bromo-3-methyl-6-1-methylethyl-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-bromo-3-methyl-6-1-methylethyl-, sodium salt 1:1, bromthymol blue sodium salt, bromothymol blue, sodium salt
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
IUPAC Name sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
Molecular Weight (g/mol) 646.37
PubChem CID 102183223
Formula Weight 646.4
Physical Form Powder
Chemical Name or Material Bromothymol blue sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.