missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Solution, Solstice

Product Code. 17238186
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17238186 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17238186 Supplier Fluka Supplier No. 346575L
  • ONLINE EXCLUSIVE PRICE
    1420.00 €
    1281.98 € / 5 L
    Save 138.02 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Bromothymol Blue Solution
CAS 76-59-5, 78-93-3, 64-17-5
CAS Max % <10%, ≥50%, ≤100%
Density 0.79 g/cm3
For Use With (Application) Laboratory
For Use With (Equipment) PANPEHA Indicator Paper
Physical Form Liquid
Quantity 5 L
Solution Type Mixture
Flash Point 13°C
Packaging Plastic Pail
UN Number UN1170
Synonym Ethanol solution
Solubility Information completely miscible
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.