missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Solution, Solstice

Product Code. 15623860
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623860 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15623860 Supplier Honeywell-Fluka Supplier No. 32822100ML
  • ONLINE EXCLUSIVE PRICE
    76.00 €
    71.40 € / 100mL
    Save 4.60 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 76-59-5
For Use With (Application) pH Calibration
Physical Form Liquid
Quantity 100 mL
Solution Type Buffer
Flash Point 38°C (100.4°F)
Molecular Formula C27H28Br2O5S
MDL Number MFCD00005872
UN Number NONH for all modes of transport
Beilstein 373934
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Molecular Weight (g/mol) 624.384
PubChem CID 6450
ChEBI CHEBI:86155
Formula Weight 624.38g/mol
Grade Not Available
Concentration or Composition (by Analyte or Components) Water (7732-18-5): ≥70.00 - <90.00 %
Ethanol (64-17-5): ≥10.00 - <20.00 %
Chemical Name or Material Bromothymol Blue Solution
Show More Show Less
compliance-icons
Product Identifier
  • Bromothymol Blue solution
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.