missing translation for 'onlineSavingsMsg'
Learn More

Bronopol, Dr. Ehrenstorfer

Product Code. 30326634
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326634 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326634 Supplier Dr. Ehrenstorfer Supplier No. DREC10810000100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bronopol
CAS 52-51-7
Analyte or Component Names Bronopol
Shipping Condition +20°C
Molecular Formula C3 H6 Br N O4
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C3H6BrNO4/c4-3(1-6,2-7)5(8)9/h6-7H,1-2H2
SMILES OCC(Br)(CO)[N+](=O)[O-]
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
Molecular Weight (g/mol) 199.99
Quantity 100 mg
Formula Weight 198.948 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.