missing translation for 'onlineSavingsMsg'
Learn More

Steris™ Browne™ Control™ Autoclave Indicator Valves

Product Code. 12705335
Change view
Click to view available options
Color:
Black Spot
Green Spot
White Spot
Yellow Spot
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12705335 Black Spot Pack of 100
12715335 Yellow Spot Pack of 100
12725335 Green Spot Pack of 100
12745335 White Spot Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12705335 Supplier Steris™ Supplier No. 7301AB
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.

  • The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
  • Various colors
  • Glass tubes
TRUSTED_SUSTAINABILITY

Specifications

Color Black Spot
For Use With (Application) For monitoring porous load steam sterilization processes
For Use With (Equipment) Autoclave
Product Type Browne Control Tube
Temperature (Metric) 121°C
Quantity 100 Pk.
Sterilization Indicator Yes
Sterilization Time 15 min.
Sterilization Method Steam, Dry Heat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.