missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Brp16 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Brp16 The Brp16 Recombinant Protein Antigen is derived from E. coli. The Brp16 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | FAM203A Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4 |
| For Use With (Application) | AC |
| Gene Alias | Brain protein 16, BRP16 |
| Gene Symbol | FAM203A |
| Label Type | Unlabeled |
| Molecular Weight (g/mol) | 28 kDa |
| Product Type | Recombinant Protein Antigen |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction