missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BS-181 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
BS-181 hydrochloride is a highly selective CDK7 inhibitor with IC50 of 21 nM, and > 40-fold selective for CDK7 than CDK1, 2, 4, 5, 6, or 9.
Specifications
Specifications
| Chemical Name or Material | BS-181 hydrochloride |
| Physical Form | Solid |
| CAS | 1397219-81-6 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H33ClN6 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | H2O : 100 mg/mL (239.81 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (119.91 mM) |
| SMILES | CC(C1=C(N=C(C=C2NCC3=CC=CC=C3)NCCCCCCN)N2N=C1)C.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur