Learn More
Novus Biologicals™ BST2 Antibody (SBI Biotech patent anti-BST2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28007-1mg
 View more versions of this product
Product Code. 30046721
  • 920.00 € / 1mg
Description
BST2 Monoclonal antibody specifically detects BST2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BST2
Monoclonal
Unconjugated
Q10589
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
SBI Biotech patent anti-BST2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN
BST2/ CD317
1 mg
Signal Transduction
684
Human
Purified