Learn More
BST2 Rabbit, anti-Human, Clone: 7J24, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82733-7-RR-20UL
This item is not returnable.
View return policy
Description
The BST2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human BST2. This antibody recognizes human antigen. The BST2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BST2 | |
| 7J24 | |
| Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin | |
| 20 μL | |
| 684 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |