missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BTBD7 Antibody Blocking Peptide

Product Code. 18504924 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18504924 0.25 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18504924 Supplier Novus Biologicals™ Supplier No. NBP149652PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 55727
Species Human
Purity >85%, by HPLC
Concentration 2.5 mg/mL
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for Antibody NBP1-49652 Applications: Immmunodepletion, Competition Assay
Gene Alias BTB (POZ) domain containing 7, DKFZp686N0544, FLJ10648, FUP1BTB/POZ domain-containing protein 7, KIAA1525, MGC48310
Gene Symbol BTBD7
Quantity 0.25 mg
Research Category Cancer
Storage Buffer Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.