missing translation for 'onlineSavingsMsg'
Learn More

BTBD9 Antibody (BTBD9/7501), Novus Biologicals™

Mouse Monoclonal Antibody

232.00 € - 462.00 €

Specifications

Antigen BTBD9
Clone BTBD9/7501
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18648644
View Documents
Novus Biologicals
NBP3-23593-100ug
100 μg
462.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18605724
View Documents
Novus Biologicals
NBP3-23593-20ug
20 μg
232.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BTBD9 Monoclonal antibody specifically detects BTBD9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

BTBD9
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
BTB (POZ) domain containing 9, BTB/POZ domain-containing protein 9, FLJ32945, KIAA1880dJ322I12.1, MGC120517, MGC120519, MGC120520
Recombinant fragment (around aa200-400) of human BTBD9 protein (exact sequence is proprietary)
Primary
Store at 4°C.
BTBD9/7501
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
114781
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.