missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTK Antibody (7F12H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
226.00 € - 473.00 €
Specifications
| Antigen | BTK |
|---|---|
| Clone | 7F12H4 |
| Concentration | 2.2 mg/ml |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18284360
|
Novus Biologicals
NB110-87047SS |
0.025 mL |
226.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795303
|
Novus Biologicals
NB110-87047 |
473.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Beschreibung
BTK Monoclonal specifically detects BTK in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| BTK | |
| 2.2 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Immunology, Innate Immunity | |
| Q06187 | |
| 695 | |
| Purified recombinant C terminal fragment of BTK expressed in E. coli. [UniProt# Q06187] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 7F12H4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human, Primate | |
| Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
| BTK | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 77 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts