missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTK, p Tyr551 Antibody (SR2188), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21550-50ul
This item is not returnable.
View return policy
Description
BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| BTK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2188 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
| A synthesized peptide derived from human phospho-BTK [Tyr551] (Uniprot #: Q06187) | |
| 50 μg | |
| Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
| 695 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction