missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA Monoclonal Antibody (OTI1E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812587
This item is not returnable.
View return policy
Description
BTLA or B and T-lymphocyte attenuator is a member of the co-inhibitory receptors of the CD28 superfamily. BTLA is a lymphocyte inhibitory receptor which inhibits lymphocytes during immunes responses. BTLA is constitutively expressed on most CD4+ and CD8+ T cells and its expression progressively decreases upon T cell activation. It remains expressed on Th1 cells, but not Th2 cells. BTLA is a unique co-receptor that interacts with the tumor necrosis factor receptor superfamily member herpesvirus entry mediator (HVEM). This interation is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response.Specifications
| BTLA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z6A9 | |
| BTLA | |
| Human recombit protein fragment corresponding to amino acids 31-157+Mouse Fc of human BTLA produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| OTI1E5 | |
| Unconjugated | |
| BTLA | |
| BTLA1, CD272 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 151888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction