missing translation for 'onlineSavingsMsg'
Learn More

BTS, MedChemExpress

Product Code. 16451816
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451816 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451816 Supplier Medchem Express Supplier No. HY16690
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BTS (N-Benzyl-p-toluenesulfonamide) is a potent and selective inhibitor of skeletal muscle myosin II subfragment 1 (S1) ATPase activity, with an IC50s of ∼5 μM for actin- and Ca2+-stimulated myosin S1 ATPase. BTS specifically inhibits the contraction of fast skeletal muscle fibers.

BTS (N-Benzyl-p-toluenesulfonamide) is a potent and selective inhibitor of skeletal muscle myosin II subfragment 1 (S1) ATPase activity, with an IC50s of ∼5 μM for actin- and Ca2+-stimulated myosin S1 ATPase. BTS specifically inhibits the contraction of fast skeletal muscle fibers.

Specifications

Chemical Name or Material BTS
CAS 1576-37-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H15NO2S
Quantity 500 mg
Synonym N-Benzyl-p-toluenesulfonamide N-Tosylbenzylamine
Solubility Information DMSO : ≥ 33.33 mg/mL (127.54 mM) ∣H2O : 0.1 mg/mL (0.38 mM; Need ultrasonic)
SMILES O=S(NCC1=CC=CC=C1)(C2=CC=C(C)C=C2)=O
Molecular Weight (g/mol) 261.34
Formula Weight 261.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.