missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody (5A9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50050G
This item is not returnable.
View return policy
Description
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry Free-Floating
Specifications
| Calbindin D-28K | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 5A9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating | |
| CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
| Full-length recombinant human Calbindin D-28K. [UniProt# P05937] | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 793 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction