missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate, Honeywell Fluka™

Product Code. 15652230 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15652230 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652230 Supplier Honeywell Fluka Supplier No. 223581500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pH 7.00

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 64-02-8, 1310-73-2, 7778-77-0, 7732-18-5
Boiling Point 100°C
CAS Min % ≥1.00%, ≥10.00%, ≥70.00%
CAS Max % <5.00%, <20.00%, <90.00%
Physical Form Liquid
Quantity 500 mL
Purity Grade Notes Buffer concentrates
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.