missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer concentrate pH 4.00, For 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer concentrate |
| pH | 4 |
| Quantity | 1 Pc. |
| Packaging | Ampoule |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?