missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate pH 4.00, For 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution, Solstice

Product Code. 15695320
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15695320 6 x 1 Ea. Pack of 6
15685320 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15695320 Supplier Fluka Supplier No. 387436X1EA
  • ONLINE EXCLUSIVE PRICE
    176.00 €
    89.07 € / Pack of 6
    Save 86.93 €
    49% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate pH 4.00
Name Note For 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution
CAS 18996-35-5, 7647-14-5
CAS Max % ≤10.0000%, ≤5.0000%
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4
Physical Form Liquid
Quantity 6 x 1 Ea.
Solution Type Buffer
Packaging Ampoule
UN Number NONH for all modes of transport
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.