missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate pH 5.00, For 500 ml buffer solution, citric acid / sodium hydroxide solution, Honeywell Fluka™

Product Code. 15615330 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
6 x 1 Ea.
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15615330 6 x 1 Ea. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615330 Supplier Honeywell-Fluka Supplier No. 387446X1EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for 500 ml buffer solution, citric acid / sodium hydroxide solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate pH 5.00
Name Note For 500 ml buffer solution, citric acid / sodium hydroxide solution
CAS 100-56-1, 7732-18-5, 1310-73-2, 5949-29-1
CAS Max % ≤0.0005%, ≤75.0000%
CAS Min % ≥69.5000%
Quantity 6 x 1 Ea.
Solution Type Buffer
MDL Number MFCD00147223
Packaging Ampoule
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.