missing translation for 'onlineSavingsMsg'
Learn More

Buffer Concentrated Solution pH10 (Borate), Fisher Chemical

Product Code. 10132050 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
6 Ampules
Unit Size:
6 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10132050 6 Ampules 6 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132050 Supplier Fisher Chemical Supplier No. J/2880C/90
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
pH 10
Physical Form Liquid
Quantity 6 Ampules
MDL Number 217672
Packaging Ampoule
compliance-icons
Product Identifier
  • Buffer solution pH 10 (borate) Concentrated solution, Traceable to NIST
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide, Boric acid (H3BO3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.