missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical

Product Code. 31751082
Change view
Click to view available options
Quantity:
4 L
Packaging:
Cubitainer
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31751082 4 L Cubitainer 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31751082 Supplier Ricca Chemical Company Supplier No. R16100004F
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7558-79-4

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 11.00± 0.01 at 25°C
CAS 7558-79-4, 1310-73-2, 7732-18-5
Melting Point 0°C
Boiling Point 100°C
CAS Min % 97.0, 1.02, 0.11
CAS Max % 100.0, 1.06, 0.11
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 11
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Packaging Cubitainer
Sterility Non-sterile
Solubility Information Miscible
Grade Reference Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.