missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Duracal pH 9.21 (25°C),NIST/PTB reference materials, DKD certified, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer Solution Duracal Ph 9.21 (25°C) |
| Name Note | Certified buffer solution traceable to NIST/PTB reference materials, DKD certified |
| Color | Blue |
| Quantity | 250 mL |
| UN Number | NONH for all modes of transport |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction