missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
6 x 1 L
Unit Size:
100mL
500mL
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 100°C |
| CAS Max % | <0.1000%, ≤0.5000% |
| CAS Min % | ≥0.2500% |
| Quantity | 6 x 1 L |
| Solution Type | Buffer |
| Packaging | Poly Bottle |
| Chemical Name or Material | Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction