missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Reagecon™
BIOPHARMA

Product Code. 11496424 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
pH:
10.0 at 20°C
10.0 at 25°C
Unit Size:
1 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
11496424 500 mL 10.0 at 25°C 500mL
11422024 500 mL 10.0 at 20°C 500mL
11412024 1000 mL 10.0 at 20°C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496424 Supplier Reagecon Supplier No. 1100525

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 10.00 is tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 10.0 at 25°C
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Odor Characteristic
Traceability to NIST Yes
Solubility Information Fully miscible in water
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 25°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.