missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10, Mettler Toledo™

Product Code. 10684863
Change view
Click to view available options
Quantity:
250 mL
Packaging:
1 Pk.
6 Pk.
Unit Size:
1500mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10684863 250 mL 1 Pk. 250mL
10422701 250 mL 6 Pk. 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10684863 Supplier Mettler Toledo Online Supplier No. 51350038
  • ONLINE EXCLUSIVE PRICE
    49.00 €
    42.51 € / 250mL
    Save 6.49 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
pH 10
Physical Form Liquid
Quantity 250 mL
Solution Type pH Buffer
Formulation 95 to 99% Deionized water, 0.1 to 0.5% Sodium hydroxide; caustic soda
Odor None
Packaging 1 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.