Learn More
Description
Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 2.00 is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Colorless |
| pH | 2.00 ±0.02 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Formulation | Glycine, Hydrochloric Acid, Sodium Chloride |
| Traceability to NIST | Yes |
| Solubility Information | Fully miscible in water |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
