missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4.00 (20°C), Solstice

Product Code. 16212892 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16212892 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16212892 Supplier Honeywell-Fluka Supplier No. 35080500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified, colored red

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution pH 4.00 (20°C)
CAS 18996-35-5, 77-92-9, 144-33-2, 7732-18-5
Melting Point <0°C
Boiling Point 100°C (1.013 hPa)
Color Red
pH 4
Quantity 500 mL
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.