missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.00, Reagecon™
BIOPHARMA

Product Code. 11416404 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
pH:
4.00 at 20°C
4.00 at 25°C
Unit Size:
1 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
11416404 500 mL 4.00 at 25°C 500mL
11422014 500 mL 4.00 at 20°C 500mL
11412014 1000 mL 4.00 at 20°C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416404 Supplier Reagecon Supplier No. 1040525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Certified by an ISO 17025 accredited test method and NIST traceable. Reagecon™ Buffer Solution, pH 4.00 is tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
pH 4.00 at 25°C
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Odor Characteristic
Traceability to NIST Yes
Solubility Information Fully miscible in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.